Stampa…

2-(4-Nitrophenyl)acetohydrazide

Fornitore: Apollo Scientific

OR111618-10G OR111618-1G OR111618-5G
APOSOR111618-10GEA 698 EUR
APOSOR111618-10G APOSOR111618-1G APOSOR111618-5G
2-(4-Nitrophenyl)acetohydrazide
2-(4-Nitrophenyl)acetohydrazide
Formula: C₈H₉N₃O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01001352
CAS Number: 6144-81-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR