Stampa…

1H-Indole-3-carbonyl chloride

Fornitore: Apollo Scientific

OR111176-1G OR111176-5G
APOSOR111176-1GEA 0 EUR
APOSOR111176-1G APOSOR111176-5G
1H-Indole-3-carbonyl chloride
1H-Indole-3-carbonyl chloride
Formula: C₉H₆ClNO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09833817
CAS Number: 59496-25-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR