3,5-Dibromo-4-methylbenzoic acid
Fornitore: Apollo Scientific
|
Attenzione
|
OR11090-25G
OR11090-5G
APOSOR11090-25GEA
215
EUR
APOSOR11090-25G
APOSOR11090-5G
3,5-Dibromo-4-methylbenzoic acid
3,5-Dibromo-4-methylbenzoic acid
Formula:
C₈H₆Br₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00017544 CAS Number: 67973-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...