4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid
Fornitore: Apollo Scientific
|
Attenzione
|
OR110759-1G
OR110759-250MG
APOSOR110759-1GEA
321
EUR
APOSOR110759-1G
APOSOR110759-250MG
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid
Formula:
C₁₀H₉NO₄ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11553038 CAS Number: 674304-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...