4-(Morpholin-4-ylmethyl)benzonitrile
Fornitore: Apollo Scientific
Synonyms:
4-(Morpholinomethyl)benzonitrile
OR110690-10G
OR110690-1G
OR110690-5G
APOSOR110690-10GEA
326
EUR
APOSOR110690-10G
APOSOR110690-1G
APOSOR110690-5G
4-(Morpholin-4-ylmethyl)benzonitrile
4-(Morpholin-4-ylmethyl)benzonitrile
Formula:
C₁₂H₁₄N₂O Peso molecolare: 202,2500 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00454271 CAS Number: 37812-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...