Stampa…

Ethyl-3-Amino-4-methylbenzoate

Fornitore: Apollo Scientific

Synonyms: 3-Amino-p-toluic Acid Ethyl Ester, Ethyl 3-amino-4-methylbenzoate, Ethyl 3-Amino-p-toluate, 3-Amino-4-methylbenzoic Acid Ethyl Ester

OR110368-10G OR110368-1G OR110368-5G
APOSOR110368-10GEA 0 EUR
APOSOR110368-10G APOSOR110368-1G APOSOR110368-5G
Ethyl-3-Amino-4-methylbenzoate
Ethyl-3-Amino-4-methylbenzoate
Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,22 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01926538
CAS Number: 41191-92-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR