Methyl-2-aminonicotinate
Fornitore: Apollo Scientific
Synonyms:
Methyl 2-aminonicotinate
OR110267-5G
OR110267-100G
OR110267-25G
APOSOR110267-5GEA
61.1
EUR
APOSOR110267-5G
APOSOR110267-100G
APOSOR110267-25G
Methyl-2-aminonicotinate
Methyl-2-aminonicotinate
Formula:
C₇H₈N₂O₂ Peso molecolare: 152,15 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00128872 CAS Number: 14667-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...