4-[(Dimethylcarbamoyl)amino]benzoic acid
Fornitore: Apollo Scientific
|
Attenzione
|
OR10864-1G
OR10864-500MG
APOSOR10864-1GEA
190
EUR
APOSOR10864-1G
APOSOR10864-500MG
4-[(Dimethylcarbamoyl)amino]benzoic acid
4-[(Dimethylcarbamoyl)amino]benzoic acid
Formula:
C₁₀H₁₂N₂O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03411197 CAS Number: 91880-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...