1-Butylboronic acid 96%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
Butylboronic acid, n-Butylboronic acid
OR10619-25G
OR10619-100G
OR10619-5G
APOSOR10619-25GEA
120
EUR
APOSOR10619-25G
APOSOR10619-100G
APOSOR10619-5G
1-Butylboronic acid 96%
1-Butylboronic acid
Formula:
C₄H₁₁BO₂ Peso molecolare: 101,94 g/mol Punto di fusione: 90…92 °C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00002106 CAS Number: 4426-47-5 EINECS: 224-607-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...