1-(3-Chlorophenyl)biguanide hydrochloride
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
m-CPBG hydrochloride
OR0928-25G
OR0928-5G
APOSOR0928-25GEA
289
EUR
APOSOR0928-25G
APOSOR0928-5G
1-(3-Chlorophenyl)biguanide hydrochloride
1-(3-Chlorophenyl)biguanide hydrochloride
Formula:
C₈H₁₁Cl₂N₅ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00053019 CAS Number: 2113-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...