Stampa…

3-Phenoxybenzonitrile 98%

Fornitore: Apollo Scientific

OR0826-1G OR0826-5G
APOSOR0826-1GEA 65.5 EUR
APOSOR0826-1G APOSOR0826-5G
3-Phenoxybenzonitrile 98%
3-Phenoxybenzonitrile
Formula: C₁₃H₉NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05669318
CAS Number: 50789-45-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR