Stampa…

Lanthanum(III) hydroxide 99.99%

Fornitore: Apollo Scientific
Pericolo

IN2140-250G IN2140-50G
APOSIN2140-250GEA 217 EUR
APOSIN2140-250G APOSIN2140-50G
Lanthanum(III) hydroxide 99.99%
Lanthanum(III) hydroxide
Formula: H₃LaO₃
Peso molecolare: 189,93 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016160
CAS Number: 14507-19-8
UN: 3262
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR