Virginiamycin M1
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
Staphylomycin, Mikamycin A
BIV1003-10MG
BIV1003-25MG
APOSBIV1003-10MGEA
109
EUR
APOSBIV1003-10MG
APOSBIV1003-25MG
Virginiamycin M1
Virginiamycin M1
Formula:
C₂₈H₃₅N₃O₇ Peso molecolare: 525,6 g/mol Temperatura di stoccaggio: Refrigerata |
CAS Number:
21411-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...