Tunicamycin (da Streptomyces species)
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₃₉H₆₄N₄O₁₆ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00065709 CAS Number: 11089-65-9 UN: 3462 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...