Stampa…

2,3-O-Isopropylidene-α,β-D-ribofuranose

Fornitore: Apollo Scientific

Synonyms: (4R,5R)-5-((R)-1,2-Dihydroxyethyl)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde

BISY003-1G BISY003-5G BISY003-10G BISY003-25G
APOSBISY003-1GEA 94.5 EUR
APOSBISY003-1G APOSBISY003-5G APOSBISY003-10G APOSBISY003-25G
2,3-O-Isopropylidene-α,β-D-ribofuranose
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose
Formula: C₈H₁₄O₅
Peso molecolare: 190,20 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01075718
CAS Number: 13199-25-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR