Stampa…

Ferulic acid methyl ester

Fornitore: Apollo Scientific

Synonyms: Methyl 4-hydroxy-3-methoxycinnamate, Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate

BISN0164-25MG BISN0164-100MG
APOSBISN0164-25MGEA 58.2 EUR
APOSBISN0164-25MG APOSBISN0164-100MG
Ferulic acid methyl ester
Ferulic acid methyl ester

MAPK/ERK Signalling Pathway

Formula: C₁₁H₁₂O₄
Peso molecolare: 208,22 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00017208
CAS Number: 2309-07-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR