Stampa…

RSL3

Fornitore: Apollo Scientific

Synonyms: (1S,3R)-2-(2-Chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylic acid methyl ester, (1S,3R)-Methyl 2-(2-chloroacetyl)-1-(4-(methoxycarbonyl)phenyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate

BISN0144-2MG BISN0144-5MG BISN0144-10MG
APOSBISN0144-2MGEA 94.5 EUR
APOSBISN0144-2MG APOSBISN0144-5MG APOSBISN0144-10MG
RSL3
RSL3

RSL3 is a ferroptosis activator in a VDAC-independent manner exhibiting selectivity for tumor cells bearing oncogenic RAS RSL3 binds inactivates GPX4 and thus mediates GPX4-regulated ferroptosis.

Formula: C₂₃H₂₁ClN₂O₅
Peso molecolare: 440,88 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD30187526
CAS Number: 1219810-16-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR