Stampa…

Paclitaxel (from Taxus Mairei)

Fornitore: Apollo Scientific
Pericolo

BIP1002-250MG BIP1002-500MG
APOSBIP1002-250MGEA 152 EUR
APOSBIP1002-250MG APOSBIP1002-500MG
Paclitaxel (from Taxus Mairei)
Paclitaxel

Potential anti-neoplastic

Formula: C₄₇H₅₁NO₁₄
Peso molecolare: 853,91 g/mol
Punto di fusione: 213 °C (dec.)
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00869953
CAS Number: 33069-62-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR