Stampa…

Oligomycin (da Streptomyces diastatochromogenes)

Fornitore: Apollo Scientific
Pericolo

BIO1002-10MG BIO1002-5MG
APOSBIO1002-10MGEA 284 EUR
APOSBIO1002-10MG APOSBIO1002-5MG
Oligomycin (da Streptomyces diastatochromogenes)
Oligomycin

Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase. A mixture of A, B, and C isomers.

Formula: C₄₅H₇₄O₁₁
Punto di fusione: 150 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD01779388
CAS Number: 1404-19-9
UN: 2811
ADR: 6.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR