Stampa…

Miconazole nitrate 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate

BIM0132-5G BIM0132-1G BIM0132-25G
APOSBIM0132-5GEA 107 EUR
APOSBIM0132-5G APOSBIM0132-1G APOSBIM0132-25G
Miconazole nitrate 98%
Miconazole nitrate

Miconazole is an imidazole antifungal agent which interferes with ergosterol synthesis and thereforealters the permeability of the cell membrane of sensitive fungi and yeasts.


  • Cell culture tested

Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
Peso molecolare: 479,14 g/mol
Punto di fusione: 184,7 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00058161
CAS Number: 22832-87-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR