Stampa…

Heparin sodium salt (from porcine intestinal mucosa)

Fornitore: Apollo Scientific
Attenzione

Synonyms: Reviparin sodium

BIH0145-250MG BIH0145-500MG BIH0145-1G
APOSBIH0145-250MGEA 50.9 EUR
APOSBIH0145-250MG APOSBIH0145-500MG APOSBIH0145-1G
Heparin sodium salt (from porcine intestinal mucosa)
Heparin sodium salt

  • min. activity >150 I.U./mg

Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00081689
CAS Number: 9041-08-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR