Stampa…

PP1 free base ≥99%

Fornitore: Apollo Scientific

Synonyms: 1-(tert-Butyl)-3-(p-tolyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

BIFK0052-10MG BIFK0052-5MG BIFK0052-1MG
APOSBIFK0052-10MGEA 176 EUR
APOSBIFK0052-10MG APOSBIFK0052-5MG APOSBIFK0052-1MG
PP1 free base ≥99%
PP1
Formula: C₁₆H₁₉N₅
Peso molecolare: 281,4 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD01076570
CAS Number: 172889-26-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR