CHIR99021 free base ≥99%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₂₂H₁₈Cl₂N₈ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD11846251 CAS Number: 252917-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...