Vatalanib free base ≥99%
Fornitore: Apollo Scientific
Synonyms:
N-(4-Chlorophenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine
BIFK0006-5MG
BIFK0006-10MG
BIFK0006-25MG
APOSBIFK0006-5MGEA
72.7
EUR
APOSBIFK0006-5MG
APOSBIFK0006-10MG
APOSBIFK0006-25MG
Vatalanib free base ≥99%
Vatalanib
Formula:
C₂₀H₁₅ClN₄ Peso molecolare: 346,8129 g/mol Punto di fusione: 204…222 °C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD08458963 CAS Number: 212141-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...