Stampa…

Dihydroergocristine methanesulphonate

Fornitore: Apollo Scientific

Synonyms: Dihydroergocristine mesylate

BID1021-1MG BID1021-5MG
APOSBID1021-1MGEA 94.5 EUR
APOSBID1021-1MG APOSBID1021-5MG
Dihydroergocristine methanesulphonate
Dihydroergocristine methanesulphonate
Formula: C₃₆H₄₅N₅O₈S
Peso molecolare: 707,843 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00153792
CAS Number: 24730-10-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR