Stampa…

X-β-D-cellobioside

Fornitore: Apollo Scientific
Attenzione

Synonyms: 5-Bromo-4-chloro-3-indolyl-β-D-cellobioside

BICS0106-1G BICS0106-250MG BICS0106-500MG
APOSBICS0106-1GEA 80 EUR
APOSBICS0106-1G APOSBICS0106-250MG APOSBICS0106-500MG
X-β-D-cellobioside
X-β-D-cellobioside
Formula: C₂₀H₂₅BrClNO₁₁
Peso molecolare: 570,78 g/mol
Temperatura di stoccaggio: Congelamento
CAS Number: 177966-52-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR