Stampa…

4-Methoxyphenyl-β-D-galactopyranoside 98% min

Fornitore: Apollo Scientific

Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol

BICL4165-10G BICL4165-25G BICL4165-5G
APOSBICL4165-10GEA 603 EUR
APOSBICL4165-10G APOSBICL4165-25G APOSBICL4165-5G
4-Methoxyphenyl-β-D-galactopyranoside 98% min
4-Methoxyphenyl-beta-D-galactopyranoside
Formula: C₁₃H₁₈O₇
Peso molecolare: 286,28 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06797126
CAS Number: 3150-20-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR