Calcitriol
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
1,25-Dihydroxycholecalciferol, (1R,3S,Z)-5-(2-((1R,3aS,7aR,E)-1-((R)-6-Hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha,25-Dihydroxyvitamin D3
BIC3222-10MG
BIC3222-25MG
BIC3222-5MG
APOSBIC3222-10MGEA
350
EUR
APOSBIC3222-10MG
APOSBIC3222-25MG
APOSBIC3222-5MG
Calcitriol
Calcitriol
Formula:
C₂₇H₄₄O₃ Peso molecolare: 416,646 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00867079 CAS Number: 32222-06-3 EINECS: 250-963-8 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...