Carbamazepine
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
5H-Dibenzo[b,f]azepine-5-carboxamide
BIC0130-100G
BIC0130-250G
BIC0130-25G
BIC0130-2KG
APOSBIC0130-100GEA
198
EUR
APOSBIC0130-100G
APOSBIC0130-250G
APOSBIC0130-25G
APOSBIC0130-2KG
Carbamazepine
Carbamazepine
Formula:
C₁₅H₁₂N₂O Peso molecolare: 236,27 g/mol Punto di fusione: 191…192 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00005073 CAS Number: 298-46-4 EINECS: 206-062-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...