Stampa…
Semicarbazide Idrocloruro
Fornitore: Apollo Scientific
Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride
BIB6269-1KG BIB6269-500G BIB6269-100G
APOSBIB6269-1KGEA 132 EUR
APOSBIB6269-1KG APOSBIB6269-500G APOSBIB6269-100G
Semicarbazide Idrocloruro
Semicarbazide Idrocloruro
Formula: CH₆ClN₃O
Peso molecolare: 111.53 g/mol
Punto di fusione: 175…177 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013009
CAS Number: 563-41-7
ADR: 6.1 III
Merck Index: 12,08588