Stampa…

Polivinilpirrolidone (K15), BioChemica

Fornitore: Apollo Scientific

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

BIA2258-50G
APOSBIA2258-50GEA 36.4 EUR
APOSBIA2258-50G
Polivinilpirrolidone (K15), BioChemica
Polyvinylpyrrolidone
Formula: (C₆H₉NO)n
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR