Fenolo equilibrated stabilizzato
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
BIA1153-250ML
APOSBIA1153-250MLEA
168
EUR
APOSBIA1153-250ML
Fenolo equilibrated stabilizzato
Fenolo
Formula:
C₆H₅OH Peso molecolare: 94,11 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 UN: 2821 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...