N-Acetyl-L-methionine ≥99%
Fornitore: Apollo Scientific
Synonyms:
N-Ac-Met-OH, Ac-Met-OH
BIA0702-100G
BIA0702-25G
APOSBIA0702-100GEA
118
EUR
APOSBIA0702-100G
APOSBIA0702-25G
N-Acetyl-L-methionine ≥99%
N-Acetyl-L-methionine
Ph. Eur.
Formula:
C₇H₁₃NO₃S Peso molecolare: 191,2 g/mol Punto di fusione: 103…106 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00064441 CAS Number: 65-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...