Stampa…

Ancymidol

Fornitore: Apollo Scientific
Attenzione

Synonyms: alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol

BIA0123-100MG BIA0123-500MG
APOSBIA0123-100MGEA 306 EUR
APOSBIA0123-100MG APOSBIA0123-500MG
Ancymidol
Ancymidol

Supports or facilitates plant growth and/or shootproliferation in two or more plant tissue cultures.

Formula: C₁₅H₁₆N₂O₂
Peso molecolare: 256,31 g/mol
Punto di fusione: 111 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00072501
CAS Number: 12771-68-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR