Stampa…

(6-Bromo-2-fluoro-3-methoxyphenyl)(methyl)sulphane 95%

Fornitore: Apollo Scientific

Synonyms: 1-Bromo-3-fluoro-4-methoxy-2-methylsulphanylbenzene

PC99012-25G PC99012-5G
APOSPC99012-25GEA 447 EUR
APOSPC99012-25G APOSPC99012-5G
(6-Bromo-2-fluoro-3-methoxyphenyl)(methyl)sulphane 95%
(6-Bromo-2-fluoro-3-methoxyphenyl)(methyl)sulphane
Formula: C₈H₈BrFOS
Peso molecolare: 251.116 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD28141768
CAS Number: 1822822-76-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR