Stampa…

Methyl (2R)-3-(4-fluorophenyl)-2-hydroxy-propanoate 95%

Fornitore: Apollo Scientific

PC911319-1G PC911319-250MG
APOSPC911319-1GEA 807 EUR
APOSPC911319-1G APOSPC911319-250MG
Methyl (2R)-3-(4-fluorophenyl)-2-hydroxy-propanoate 95%
Methyl (2R)-3-(4-fluorophenyl)-2-hydroxy-propanoate
Formula: C₁₀H₁₁FO₃
Peso molecolare: 198,191 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16495900
CAS Number: 124980-98-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR