3-Acetyl-4-fluorobenzoic acid 97%
Fornitore: Apollo Scientific
PC910947-1G
PC910947-5G
APOSPC910947-1GEA
256
EUR
APOSPC910947-1G
APOSPC910947-5G
3-Acetyl-4-fluorobenzoic acid 97%
3-Acetyl-4-fluorobenzoic acid
Formula:
C₉H₇FO₃ Peso molecolare: 182,148 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD22493249 CAS Number: 1505662-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...