Stampa…

N-Ethyl-3-fluoroaniline 95%

Fornitore: Apollo Scientific

PC910847-1G PC910847-250MG
APOSPC910847-1GEA 754 EUR
APOSPC910847-1G APOSPC910847-250MG
N-Ethyl-3-fluoroaniline 95%
N-Ethyl-3-fluoroaniline
Formula: C₈H₁₀FN
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11144550
CAS Number: 2707-62-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR