Stampa…

Methyl 3-(2-fluorophenyl)-3-oxopropanoate 95%

Fornitore: Apollo Scientific

PC907946-1G PC907946-25G PC907946-5G
APOSPC907946-1GEA 109 EUR
APOSPC907946-1G APOSPC907946-25G APOSPC907946-5G
Methyl 3-(2-fluorophenyl)-3-oxopropanoate 95%
Methyl 3-(2-fluorophenyl)-3-oxopropanoate
Formula: C₁₀H₉FO₃
Peso molecolare: 196,175 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03424763
CAS Number: 185302-86-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR