Stampa…

Methyl 5-(trifluoromethyl)pyrazole-3-carboxylate 98%

Fornitore: Apollo Scientific

PC904751-1G PC904751-250MG PC904751-5G
APOSPC904751-1GEA 343 EUR
APOSPC904751-1G APOSPC904751-250MG APOSPC904751-5G
Methyl 5-(trifluoromethyl)pyrazole-3-carboxylate 98%
Methyl 5-(trifluoromethyl)pyrazole-3-carboxylate
Formula: C₆H₅F₃N₂O₂
Peso molecolare: 194,111 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09754160
CAS Number: 6833-82-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR