Benzyl 3-fluoropropanoate 95%
Fornitore: Apollo Scientific
Formula:
C₁₀H₁₁FO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD21332902 CAS Number: 115952-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...