Stampa…

1-Bromo-4-methoxy-2-(trifluoromethoxy)benzene 95%

Fornitore: Apollo Scientific

Synonyms: 4-Methoxy-2-(trifluoromethoxy)bromobenzene

PC903007-1G PC903007-25G PC903007-5G
APOSPC903007-1GEA 65.4 EUR
APOSPC903007-1G APOSPC903007-25G APOSPC903007-5G
1-Bromo-4-methoxy-2-(trifluoromethoxy)benzene 95%
1-Bromo-4-methoxy-2-(trifluoromethoxy)benzene
Formula: C₈H₆BrF₃O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09258678
CAS Number: 1049730-84-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR