2-Fluoro-N-methyl-6-nitroaniline 98%
Fornitore: Apollo Scientific
PC902097-10G
PC902097-1G
PC902097-5G
APOSPC902097-10GEA
502
EUR
APOSPC902097-10G
APOSPC902097-1G
APOSPC902097-5G
2-Fluoro-N-methyl-6-nitroaniline 98%
2-Fluoro-N-methyl-6-nitroaniline
Formula:
C₇H₇FN₂O₂ Peso molecolare: 170,141 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD23725947 CAS Number: 182551-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...