Stampa…

1-(4-Fluorophenyl)-2,5-dimethylpyrrole 97%

Fornitore: Apollo Scientific

PC902045-1G PC902045-5G PC902045-10G
APOSPC902045-1GEA 168 EUR
APOSPC902045-1G APOSPC902045-5G APOSPC902045-10G
1-(4-Fluorophenyl)-2,5-dimethylpyrrole 97%
1-(4-Fluorophenyl)-2,5-dimethylpyrrole
Formula: C₁₂H₁₂FN
Peso molecolare: 189,229 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01910971
CAS Number: 54609-08-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR