1-Chloro-2-nitro-4-(trifluoromethoxy)benzene
Fornitore: Apollo Scientific
Formula:
C₇H₃ClF₃NO₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD13194447 CAS Number: 588-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...