Stampa…

(R)-2-Fluoromandelic acid 98%

Fornitore: Apollo Scientific

PC900788-10G PC900788-1G PC900788-5G
APOSPC900788-10GEA 574 EUR
APOSPC900788-10G APOSPC900788-1G APOSPC900788-5G
(R)-2-Fluoromandelic acid 98%
(R)-2-Fluoromandelic acid
Formula: C₈H₇FO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04038772
CAS Number: 32222-48-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR