Stampa…

Sodium difluoromethanesulphinate 95%

Fornitore: Apollo Scientific

PC900211-1G PC900211-250MG
APOSPC900211-1GEA 742 EUR
APOSPC900211-1G APOSPC900211-250MG
Sodium difluoromethanesulphinate 95%
Sodium difluoromethanesulphinate
Formula: CHF₂O₂S·Na
Peso molecolare: 138,069 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD24113355
CAS Number: 275818-95-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR