5-Fluoro-2-isopropoxybenzonitrile 96%
Fornitore: Apollo Scientific
Formula:
C₁₀H₁₀FNO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD15201676 CAS Number: 1158506-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...