Stampa…

N-Methyl-2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine 97%

Fornitore: Apollo Scientific

PC8547-1G PC8547-250MG
APOSPC8547-1GEA 230 EUR
APOSPC8547-1G APOSPC8547-250MG
N-Methyl-2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine 97%
N-Methyl-2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine
Formula: C₁₃H₁₄F₃N₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09817515
CAS Number: 898598-62-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR