Stampa…

3,5-Dimethyl-2-fluoropyridine 95+%

Fornitore: Apollo Scientific

Synonyms: 2-Fluoro-3,5-dimethylpyridine

PC6839-1G PC6839-250MG PC6839-5G
APOSPC6839-1GEA 440 EUR
APOSPC6839-1G APOSPC6839-250MG APOSPC6839-5G
3,5-Dimethyl-2-fluoropyridine 95+%
3,5-Dimethyl-2-fluoropyridine
Formula: C₇H₈FN
Peso molecolare: 125,144 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11044345
CAS Number: 111887-71-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR